accessibility menu, dialog, popup

Test

CAS RN 107549-22-4

CAS RN 107549-22-4

IUPAC Name: magnesium(2+) (3-bromophenyl)methanide bromide
Synonyms: magnesium(2+) (3-bromophenyl)methanide bromide
Molecular Weight (g/mol): 274.24
Molecular Formula: C7H6Br2Mg
InChi Key: OJXOOFXUHZAXLO-UHFFFAOYSA-M
SMILES: [Mg++].[Br-].[CH2-]C1=CC(Br)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results