CAS RN 106724-85-0
CAS RN 106724-85-0
IUPAC Name:
1-benzyl-3,5-dibromo-1H-1,2,4-triazole
Synonyms:
1-benzyl-3,5-dibromo-1,2,4-triazole
Molecular Weight (g/mol):
316.98
Molecular Formula:
C9H7Br2N3
InChi Key:
YXSNCHAOAYRKCF-UHFFFAOYSA-N
SMILES:
BrC1=NN(CC2=CC=CC=C2)C(Br)=N1