Learn More
Thermo Scientific™ Zeba Spin 7K MWCO Desalting Columns
Effectively remove salt and other molecules (7K MWCO) from samples and attain excellent protein recovery with these centrifuge-based chromatography columns.
59.75€ - 391.00€
Specifications
Formulation Polypropylene columns with proprietary resin
MWCO 7000 Da
View More Specs
Products 10
Product Code Brand Sufficient For Quantity Size (Metric) Price Quantity & Availability  
Product Code Brand Sufficient For Quantity Size (Metric) Price Quantity & Availability  
10678294 Thermo Scientific™
89878
50 samples, each 2 to 12uL 50 columns 75uL 273.00€
Pack of 50 View alternative products
Save Up to 
»
EU Stock 16
Add to basket
10627354 Thermo Scientific™
89890
25 samples, each 200 to 700uL 25 columns 2mL 255.00€
Pack of 25 View alternative products
Save Up to 
»
EU Stock 163
Add to basket
10657514 Thermo Scientific™
89891
5 samples, each 500 to 2000uL 5 columns 5mL 74.50€
Pack of 5 View alternative products
Save Up to 
»
EU Stock 38
Add to basket
10415545 Thermo Scientific™
89889
5 samples, each 200 to 700uL 5 columns 2mL 59.75€
Pack of 5 View alternative products
Save Up to 
»
EU Stock 87
Add to basket
10667514 Thermo Scientific™
89893
5 samples, each 700uL to 4mL 5 columns 10mL 92.25€
Pack of 5 View alternative products
Save Up to 
»
EU Stock 260
Add to basket
10435725 Thermo Scientific™
89894
25 samples, each 700uL to 4mL 25 columns 10mL 391.00€
Pack of 25 View alternative products
Save Up to 
»
EU Stock 52
Add to basket
10290384 Thermo Scientific™
89877
25 samples, each 2 to 12uL 25 columns 75uL 155.00€
Pack of 25 View alternative products
Save Up to 
»
EU Stock 33
Add to basket
10341164 Thermo Scientific™
89882
25 samples, each 30 to 130uL 25 columns 0.5mL 141.00€
Pack of 25 View alternative products
Save Up to 
»
EU Stock 215
Add to basket
10056033 Thermo Scientific™
89883
50 samples, each 30 to 130uL 50 columns 0.5mL 253.00€
Pack of 50 View alternative products
Save Up to 
»
EU Stock 107
Add to basket
10280584 Thermo Scientific™
89892
25 samples, each 500 to 2000uL 25 columns 5mL 326.00€
Pack of 25 View alternative products
Save Up to 
»
EU Stock 49
Add to basket
Description
Description
These specially designed polypropylene centrifuge columns are pre-packed with high-performance Zeba Desalting Resin to facilitate rapid protein desalting with high recovery. The resin effectively traps salts and other small sample components but does not impede proteins and other macromolecules, which pass directly into the centrifuge collection tube. These particular columns use Zeba Resin having a molecular-weight cutoff (MWCO) rating equal to 7K; this ensures that proteins larger than 7000 MW will be recovered with high efficiency.
The easy-to-use spin column format dramatically improves results over standard drip-column methodologies. This system eliminates the need to wait for samples to emerge by gravity flow and to monitor fractions for protein recovery. Samples are processed in these desalting columns using a standard centrifuge or microcentrifuge. No chromatography hardware or column preparation steps are required. Multiple samples can be processed in less than 10 minutes. Zeba Spin Desalting Columns are available in five different column sizes, and these provide trouble-free desalting and buffer exchange for sample volumes ranging from 2μL to 4mL.
Highlights:
  • Fast centrifuge format – resin and columns are optimized for rapid processing by centrifugation with standard collection tubes and rotors
  • Replaces drip-column methods – no need to collect and screen slowly emerging protein fractions
  • Exceptional protein yields – recover greater than 95% in typical applications with minimal dilution
  • Five convenient sizes – columns with 75μL to 10mL of Zeba Resin provide desalting matched for sample volumes between 2μL and 4mL
  • 7K MWCO – resin is rated to recover proteins larger than 7000 daltons and retain (remove) solutes smaller than 1000 MW
Requires:
Microcentrifuge and microcentrifuge collection tubes (for 75μL and 0.5mL columns) or centrifuge and collection tubes (15mL tubes for 2 and 5mL columns; 50mL tubes for 10mL columns)
Specifications
Specifications
Polypropylene columns with proprietary resin
7000 Da