Learn More
Buerkle™ Polystyrene Volumetric Spoons
Product Code. 10274904 Shop All Bürkle Products
Change view
Click to view available options
Capacity (Metric):
0.5 to 50 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10274904 0.5 to 50 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10274904 Supplier Buerkle™ Supplier No. 53340001
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
The capacity is embossed into the spoon handle. Available as a set with 8 volumetric spoons or separate in bags of 100 volumetric spoons.
  • Polystyrene, blue
  • Round shape, easy emptying
TRUSTED_SUSTAINABILITY
Specifications
Quantity 8 Pieces/Pk.
Capacity (Metric) 0.5 to 50 mL
Color Blue
Product Type Measuring Spoon Set
Material Polystyrene