missing translation for 'onlineSavingsMsg'
Learn More

Tricine, 98+%

Product Code. 11407706
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11407706 100 g 100g
11497696 25 g 25g
11417706 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11407706 Supplier Thermo Scientific Alfa Aesar Supplier No. A14695.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5704-04-1 | C6H13NO5 | 179.17 g/mol

Good's buffers Tricine is used as an electrophoresis buffer and involved in the separation of low molecular weight proteins and peptides. It operates in the pH range between 7.4 and 8.8. It acts as an effective scavenger of hydroxyl radicals during the study of radiation-induced membrane damage. It is also used in resuspension of cell pellets.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Good′s buffers Tricine is used as an electrophoresis buffer and involved in the separation of low molecular weight proteins and peptides. It operates in the pH range between 7.4 and 8.8. It acts as an effective scavenger of hydroxyl radicals during the study of radiation-induced membrane damage. It is also used in resuspension of cell pellets.

Solubility
Soluble in water.

Notes
pH range: 7.8-8.8Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4-1-5704
Molecular Formula C6H13NO5
Molecular Weight (g/mol) 179.17
MDL Number MFCD00004277
InChI Key SEQKRHFRPICQDD-UHFFFAOYSA-N
Synonym N-(Trishydroxymethylmethyl)glycine
PubChem CID 79784
ChEBI CHEBI:39063
IUPAC Name 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid
SMILES C(C(=O)O)NC(CO)(CO)CO

Specifications

Melting Point ∼183°C (decomposition)
Quantity 100 g
Beilstein 1937804
Merck Index 14,9651
Solubility Information Soluble in water.
Formula Weight 179.17
Percent Purity ≥98%
Chemical Name or Material Tricine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.