missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMPRSS11BNL Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA572111
Additional Details : Weight : 0.01000kg
Description
TMPRSS11BNL is a protein coding gene.Specifications
TMPRSS11BNL | |
Polyclonal | |
Unconjugated | |
TMPRSS11BNL | |
TMPRSS11B N-terminal like, pseudogene; TMPRSS11BNL | |
Rabbit | |
Protein A | |
RUO | |
401136 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.45 mg/mL | |
PBS with 0.09% sodium azide | |
0 | |
TMPRSS11BNL | |
KLH conjugated synthetic peptide between 43-69 amino acids from the Central region of human TMPRSS11BNL | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |