Learn More
Ley Rubber™ Tapered Cork Stoppers
Tapered cork stoppers for use in education, homebrewing and arts and crafts.
2.89€ - 140.00€
Specifications
Material | Cork |
---|---|
Product Type | Cork Stopper |
Shape | Tapered |
For Use With (Application) | Education, homebrewing, arts and crafts |
Product Code | Brand | Length (Metric) | Diameter (Metric) Top | Diameter (Metric) Bottom | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) Top | Diameter (Metric) Bottom | Price | Quantity & Availability | |||
12396457
|
Ley Rubber™
CB018/S/Z |
32 mm | 21 mm | 18 mm |
2.89€
Pack of 10 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||
12376457
|
Ley Rubber™
CB015/S/Z/2 |
24 mm | 18 mm | 15 mm |
4.50€
Pack of 20 |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||
12336477
|
Ley Rubber™
CB011/S/Z |
19 mm | 14 mm | 11 mm |
140.00€
Pack of 1000 |
Estimated Shipment: 22-07-2024
Log in to see stock. |
|||
Description
- Range of sizes available
- Recyclable product
Specifications
Cork | |
Cork Stopper | |
Tapered | |
Education, homebrewing, arts and crafts |