Learn More
Thermo Scientific™ Syringes for Thermo Scientific™ HPLC Instruments
Thermo Scientific™ Syringes for Thermo Scientific HPLC Instruments offer easy, accurate and reproducible injections.
180.62€ - 183.00€
Product Code | Brand | For Use With (Equipment) | Volume (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Volume (Metric) | Price | Quantity & Availability | ||||
10782127
|
Thermo Scientific™
365ILT21 |
LCQ HPLC | 250 μL |
180.62€
Each |
In Stock
Log in to see stock. |
||||
10341205
|
Thermo Scientific™
365JLT41 |
LCQ HPLC | 500 μL |
183.00€
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
||||
10702917
|
Thermo Scientific™
365ILT91 |
AS1000, AS3000 Autosampler | 250 μL |
182.98€
Each |
Estimated Shipment: 29-05-2024
Log in to see stock. |
||||
10321065
|
Thermo Scientific™
365JLT61 |
AS1000, AS3000 Autosampler | 500 μL |
182.98€
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
||||
10197744
|
Thermo Scientific™
365LLT81 |
AS3000, AS3500 Autosampler | 2.5 mL |
182.98€
Each |
Estimated Shipment: 24-06-2024
Log in to see stock. |
||||
Description
Thermo Scientific™ Syringes for Thermo Scientific HPLC Instruments offer easy, accurate and reproducible injections.
- Square tip to prevent damage to the injector
- Wide range of volumes
- Precision made from borosilicate glass and stainless steel
- Robust design and easy-to-read markings
Specifications
HPLC Autosampler Syringe | |
No |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.