Learn More
Socorex™ Dosys™ All-glass Syringes
Resistant borosilicate glass syringes can be sterilized and reused. Socorex™ Dosys™ All-glass Syringes provide a low-cost alternative to disposable plastic syringes and are easy to disassemble for cleaning.
28.35€ - 405.00€
Product Code | Brand | Volume (Metric) | Connection Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Connection Type | Price | Quantity & Availability | ||||
10477072
|
Socorex™
155.0502 |
0.5 to 2 mL | Luer Lock |
30.85€
Pack of 3 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||||
10029761
|
Socorex™
155.0520 |
1 to 20 mL | Luer Lock |
37.15€
Pack of 2 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
10131442
|
Socorex™
155.0550 |
1 to 50 mL | Luer Lock |
43.05€
Each |
In Stock
Log in to see stock. |
||||
10039701
|
Socorex™
155.0305 |
0.2 to 5 mL | Luer Nozzle |
28.35€
Pack of 3 |
Estimated Shipment: 20-06-2024
Log in to see stock. |
||||
10324932
|
Socorex™
155.0320 |
1 to 20 mL | Luer Nozzle |
33.35€
Pack of 2 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
||||
10161342
|
Socorex™
155.0310 |
1 to 10 mL | Luer Nozzle |
32.00€
Pack of 3 |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||||
10551713
|
Socorex™
155.0505 |
0.2 to 5 mL | Luer Lock |
32.00€
Pack of 3 |
Estimated Shipment: 14-06-2024
Log in to see stock. |
||||
10385022
|
Socorex™
155.0510 |
1 to 10 mL | Luer Lock |
33.65€
Pack of 3 |
Estimated Shipment: 19-06-2024
Log in to see stock. |
||||
11837253
|
Socorex™
155.0301 |
0.1 to 1 mL | Luer Nozzle |
28.35€
Pack of 3 |
Estimated Shipment: 13-06-2024
Log in to see stock. |
||||
12663275
|
Socorex™
155.05150 |
10 to 150 mL | Luer Lock |
392.00€
Each |
Estimated Shipment: 19-08-2024
Log in to see stock. |
||||
12673275
|
Socorex™
155.05200 |
10 to 200 mL | Luer Lock |
405.00€
Each |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||||
15562185
|
Socorex™
155.0330 |
1 to 30 mL | Luer Nozzle |
39.00€
Pack of 2 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||||
Description
- Borosilicate glass construction.
- Superior chemical, heat and shock resistance.
- Low-cost alternative to disposable plastic syringes.
- Precision-machined plunger and barrel.
- Excellent fitting and liquid tightness (not gas tight).
- Permanent, high visibility graduations.
- Glass Luer and metal Luer lock nozzles available.
- Can be sterilized up to 160°C / 320°F.
- Very easy to disassemble and clean.
Specifications
Autoclavable | |
Syringe | |
Yes |