Learn More
STEMCELL Technologies STEMtaq™ DNA Polymerase Master Mix Kit
DNA polymerase master mix and nuclease-free water for PCR reactions
POA
Specifications
For Use With (Application) Real Time PCR (qPCR)
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17109901
View Documents Promotion Details
STEMCELL Technologies
79009
100 Reactions
POA
1 set
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.
Request a Price for this item here.
17119901
View Documents Promotion Details
STEMCELL Technologies
79009.1
1000 Reactions
POA
1 set
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.
Request a Price for this item here.
Description
Description
  • STEMtaq™ DNA Polymerase Master Mix Kit, which includes master mix and nuclease-free water, reliably and consistently amplifies a wide range of PCR templates
  • STEMtaq™ DNA Polymerase Master Mix is a 2X concentrated, ready-to-use solution containing STEMtaq™ DNA polymerase, dNTPs, MgCl2, and reaction buffer
  • STEMtaq™ DNA polymerase has 5' to 3' exonuclease activity and produces PCR fragments with a 3' A overhang
Specifications
Specifications
Real Time PCR (qPCR)