Learn More
STEMCELL Technologies STEMtaq™ DNA Polymerase Master Mix Kit
DNA polymerase master mix and nuclease-free water for PCR reactions
POA
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17109901
|
STEMCELL Technologies
79009 |
100 Reactions |
POA
1 set |
N/A | |||||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
17119901
|
STEMCELL Technologies
79009.1 |
1000 Reactions |
POA
1 set |
N/A | |||||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
Description
- STEMtaq™ DNA Polymerase Master Mix Kit, which includes master mix and nuclease-free water, reliably and consistently amplifies a wide range of PCR templates
- STEMtaq™ DNA Polymerase Master Mix is a 2X concentrated, ready-to-use solution containing STEMtaq™ DNA polymerase, dNTPs, MgCl2, and reaction buffer
- STEMtaq™ DNA polymerase has 5' to 3' exonuclease activity and produces PCR fragments with a 3' A overhang
Specifications
Real Time PCR (qPCR) |