Learn More
Thermo Scientific™ Manual, Removable-Needle, Gas Tight Syringes for GC Instruments
Use Thermo Scientific™ Removable Needle Syringes for accurate and reproducible manual sample introduction into GC instruments, with or without the needle.
64.17€ - 152.09€
Product Code | Brand | Volume (Metric) | Needle Point Style | Needle Gauge | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Needle Point Style | Needle Gauge | Price | Quantity & Availability | |||
10111033
|
Thermo Scientific™
365DLG21 |
10 μL | 90° Blunt | 22 G |
94.23€
Each |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||
10630564
|
Thermo Scientific™
365GLG41 |
50 μL | 90° Blunt | 22 G |
85.56€
Each |
In Stock
Log in to see stock. |
|||
10523035
|
Thermo Scientific™
365HLG51 |
100 μL | 90° Blunt | 22 G |
92.64€
Each |
Estimated Shipment: 01-07-2024
Log in to see stock. |
|||
10435923
|
Thermo Scientific™
365JLG71 |
500 μL | Blunt | 22s G |
93.86€
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||
15234573
|
Thermo Scientific™
365D0811 |
10 μL | Bevel | 26s G |
64.17€
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||
15224583
|
Thermo Scientific™
365M5212 |
5 mL | Bevel | 22 G |
128.00€
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||
15244583
|
Thermo Scientific™
365N5214 |
10 mL | Bevel | 22 G |
121.20€
Each |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||
15274583
|
Thermo Scientific™
365N8923 |
10 mL | Side Hole | 22s G |
152.09€
Each |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||
10117714
|
Thermo Scientific™
365F1931 |
25 μL | Bevel | 25 G |
85.56€
Each |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||
10127744
|
Thermo Scientific™
365J2881 |
500 μL | Bevel | 25 G |
91.49€
Each |
Estimated Shipment: 19-06-2024
Log in to see stock. |
|||
10640626
|
Thermo Scientific™
365K3041 |
1 mL | Bevel | 23 G |
107.58€
Each |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||
15254583
|
Thermo Scientific™
365N6323 |
10 mL | Blunt | 22s G |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||
11524070
|
Thermo Scientific™
365KLG81 |
1 mL | Blunt | 22s G |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||
10552455
|
Thermo Scientific™
365FLG31 |
25 μL | Blunt | 22s G |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||
15224603
|
Thermo Scientific™
365M6322 |
5 mL | Blunt | 22s G |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||
10394563
|
Thermo Scientific™
365ILG61 |
250 μL | Blunt | 22 G |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||
Description
Get durability, clarity and accuracy from Thermo Scientific™ Manual Removable Needle Syringes. Used for manual sample injection into gas chromatography (GC) instruments with or without the needle, they provide accuracy and reproducibility that provides confidence in analytical results, time-after-time. Choose from a variety of sizes, and types. These syringes are available in standard industry size with black easy-to-read and solvent-resistant markings.
- Square tip to prevent damage to the injector
- Wide range of volumes
- Precision made from borosilicate glass and stainless steel
- Robust design and easy-to-read markings
Specifications
50 mm | |
1/Ea. | |
50 mm |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.