Learn More
Koettermann™ Standard Hanging Cabinet, 1500x780x366
Koettermann™ Standard Hanging Cabinet, 1500x780x366 provides suitable storage to accommodate a variety of laboratory needs.
980.93€ - 1062.97€
Specifications
Dimensions (D x W x H) | 350 x 1500 x 780 mm |
---|---|
No. of Doors | 3 |
No. of Shelves | 1 |
Product Type | Wall Mounted Cabinet |
Product Code | Brand | Locking Mechanism | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Locking Mechanism | Price | Quantity & Availability | |||||
15763122
|
Koettermann™
307-00210 |
Yes |
1062.97€
Each |
N/A | |||||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
15753122
|
Koettermann™
307-00209 |
No |
980.93€
Each |
N/A | ||||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
Description
- Wall-mounted cabinet
- Three doors
- One shelf
- Side panels
Specifications
350 x 1500 x 780 mm | |
3 | |
1 | |
Wall Mounted Cabinet |