Learn More
Thermo Scientific™ Standard Fixed Needle Syringes for Rheodyne/Valco™ Injectors
Thermo Scientific™ Standard Fixed Needle Syringes for Rheodyne/Valco Injectors offer easy, accurate and reproducible manual injections.
42.01€ - 93.86€
Specifications
Product Type | HPLC Autosampler Syringe |
---|---|
Disposable | No |
For Use With (Equipment) | Injector |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
10171073
|
Thermo Scientific™
365GL251 |
50 μL |
60.95€
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
10631714
|
Thermo Scientific™
365JL281 |
500 μL |
93.86€
Each |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
10715564
|
Thermo Scientific™
365FL241 |
25 μL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
10324133
|
Thermo Scientific™
365CL221 |
5 μL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
10160503
|
Thermo Scientific™
365HL261 |
100 μL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
10120553
|
Thermo Scientific™
365IL271 |
250 μL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
10110743
|
Thermo Scientific™
365DL231 |
10 μL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Description
Thermo Scientific™ Standard Fixed Needle Syringes for Rheodyne/Valco Injectors offer easy, accurate and reproducible manual injections.
- Square tip to prevent damage to the injector
- Wide range of volumes
- Precision made from borosilicate glass and stainless steel
- Robust design and easy-to-read markings
Specifications
HPLC Autosampler Syringe | |
No | |
Injector |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.