Learn More
Lenz Laborglasinstrumente™ Spiral Springs
32.54€
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
10221071
|
Lenz Laborglasinstrumente™
01102825 |
25 mm |
32.54€
Pack of 10 |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||||
10393131
|
Lenz Laborglasinstrumente™
01102835 |
35 mm |
32.54€
Pack of 10 |
Estimated Shipment: 02-07-2024
Log in to see stock. |
|||||