Learn More
Fisherbrand™ Spares Kit
For use with Fisherbrand™ Vacuum Pumps to aid in evacuating a 1L system to 200mbar in 40 seconds.
Brand: Fisherbrand™
Additional Details : Weight : 0.20000kg
Description
- Includes washers, O-rings and inserts
- Accessory item
Spécification
Vacuum Pump | |
Washers, O-rings and inserts | |
1Each | |
1 Each |