Learn More
Fisherbrand™ Soda Lime Glass Light Wall Rimmed Test Tube
Manufactured from soda lime glass and constructed with light walls. Fisherbrand™ Soda Lime Glass Light Wall Rimmed Test Tubes are ideal for general mixing and laboratory work.
14.90€ - 38.65€
Specifications
Certifications/Compliance | ISO 4142 |
---|---|
Material | Soda Lime Glass |
Rimmed | Yes |
Product Code | Brand | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
15832265
|
Fisherbrand™
|
12 mm | 100 mm |
18.45€
Pack of 100 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
||||
15852265
|
Fisherbrand™
|
16 mm | 125 mm |
21.00€
Pack of 100 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
||||
15842275
|
Fisherbrand™
|
24 mm | 150 mm |
38.65€
Pack of 50 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
||||
15872265
|
Fisherbrand™
|
16 mm | 150 mm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||||
15812265
|
Fisherbrand™
|
12 mm | 75 mm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||||
15892255
|
Fisherbrand™
|
10 mm | 75 mm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||||
Description
- Withstands moderate warming
- Light weight
- Complies with ISO 4142
Specifications
ISO 4142 | |
Soda Lime Glass | |
Yes |