Learn More
Fisherbrand™ Soda Glass Funnel
Use the Fisherbrand™ Soda Glass Funnel to funnel liquids for laboratory applications. the funnel is manufactured from clear soda lime glass and has a stem that measures 40mm.
Brand:  Fisherbrand™
Additional Details : Weight : 0.01800kg
Product Code. 11502423
  • 2.60€ / Each
Estimated Shipment: 11-06-2024
Description
  • Soda lime glass
  • Conical shape
  • 1Pack
Specifications
Specifications
35mm
Soda Lime Glass
40mm