Learn More
Epredia™ Shandon™ Instrument Case
Epredia™ Shandon Instrument Case is available as a complete kit or case only.
Brand:  Epredia™ 9996969
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 15338278
  • 2995.00€ / 1 set
Estimated Shipment: 17-09-2024
Description
Dimensions: 12.75L × 9.5W × 2.5in.D (32.4 × 24.1 × 6.4cm)
Specifications
Specifications
Instrument Case
Case with instruments