Learn More
Sartorius Printer Paper
Paper for various Sartorius printers.
24.30€ - 25.50€
Product Code | Brand | For Use With (Equipment) | Length (English) | Length (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Length (English) | Length (Metric) | Price | Quantity & Availability | |||
10663182
|
Sartorius
6906937 |
YDP20-0CE Printer | 1944 in. | 4937.76 cm |
24.30€
Pack of 5 |
In Stock
Log in to see stock. |
|||
11338963
|
Sartorius
6906920 |
YDP06 Dot Matrix Printer | 550 in. | 1397 cm |
25.50€
Each |
In Stock
Log in to see stock. |
|||
Description
Paper for use with balance printers.Specifications
Printer Paper |