Learn More
STEMCELL Technologies ReproRNA™-OKSGM Kit
A non-integrating, self-replicating RNA reprogramming vector for generating iPS cells
Brand:  STEMCELL Technologies 05930
Additional Details : Weight : 0.01330kg Transport : UN number : 1170 Chem class : 3 Pack group : II
Product Code. 17128311
  • 3250.50€ / 1 set
Estimated Shipment: 22-05-2024
to see stock.
Add to basket
Description
Description
  • ReproRNA™-OKSGM is a single-stranded RNA replicon vector that contains five reprogramming factors: OCT4, KLF-4, SOX2, GLIS1, and c-MYC, as well as a puromycin-resistance gene (Figure 1)
  • This RNA vector reprograms somatic cells, such as fibroblasts, into induced pluripotent stem (iPS) cells with high efficiency and only requires a single transfection step (Figure 2)
  • When used together with ReproTeSR™ reprogramming medium, the generation of iPS cell colonies can be achieved under feeder-free conditions with superior colony morphology and similar reprogramming efficiency to feeder-based systems (Figures 3 - 4)
  • ReproRNA™-derived iPS cell colonies also express markers of undifferentiated cells and retain a normal karyotype (Figures 5-6)
  • Subsequently, iPS cells generated with ReproRNA™-OKSGM can be maintained in TeSR™ maintenance media (mTeSR™1, TeSR™2, or TeSR™-E8™) and further differentiated into cells of all three germ layers (Figure 7)
Specifications
Specifications
Pluripotent Stem Cells
Cell Culture; Reprogramming
1 Kit