Learn More
Deutsch Neumann™ Red Natural Rubber Filter Ring with Flange
Brand:  Deutsch Neumann™ 2350012
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 10186590
  • 1.40€ / Each
Estimated Shipment: 07-06-2024
Specifications
Red
1Each
41 mm
Natural Rubber
1 Each
20 mm