missing translation for 'onlineSavingsMsg'
Learn More

Rat anti-Mouse IgG (H+L) Secondary Antibody, HRP, Invitrogen™

Rat Monoclonal Secondary Antibody

Brand:  Invitrogen SA135636

Additional Details : Weight : 0.01000kg

Product Code. 11555080

  • 2335.00€ / 1mg
Estimated Shipment: 04-06-2024
to see stock.
Add to basket

Description

Description

Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications

Specifications

Mouse IgG (H+L)
Monoclonal
1 mg/mL
PBS with 50% glycerol and no preservative
Immunoglobulins.
1 mg
Secondary
-20° C, Avoid Freeze/Thaw Cycles
IgM κ
ELISA, Immunohistochemistry (Frozen)
LO-MG-7
HRP
Rat
purified
RUO
Mouse
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers