Learn More
Edmund Bühler GmbH™ PVC Support Rods
Six support rods for holding six vessels. For use with IRB 6 support base.
Brand:  Edmund Bühler GmbH™ 0052 091
Additional Details : Weight : 0.25000kg
Product Code. 10408662
  • 83.10€ / Each
Estimated Shipment: 03-07-2024
Description
  • Support rods constructed of PVC
  • For use with support base IRB 6
  • Fasten clamps for 6 glass vessels
Specifications
Specifications
Tripod Stand
PVC Rail