Learn More
Eppendorf™ PCR Tubes - Singles and Strips
GREENER_CHOICE
Product Code. p-8003117 Shop All Eppendorf Products
Change view
Click to view available options
Closure Type:
Domed Strip Cap
Flat Strip Cap
Hinged Cap
Not Included
Format:
8-tube Strips
Fast PCR Tube Strips
Single Tube
Purity or Quality Grade:
Forensic DNA Grade
PCR Clean (Non-sterile)
Volume (Metric):
0.1 mL
0.2 mL
0.5 mL
Unit Size:
Pack of 1000
Pack of 120
Pack of 500
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Closure Type Format Purity or Quality Grade Volume (Metric) unitSize
15645367 Hinged Cap Single Tube Forensic DNA Grade 0.2 mL Pack of 500
10500082 Hinged Cap Single Tube PCR Clean (Non-sterile) 0.2 mL Pack of 1000
10401203 Hinged Cap Single Tube PCR Clean (Non-sterile) 0.5 mL Pack of 500
11843432 Not Included 8-tube Strips PCR Clean (Non-sterile) 0.1 mL Pack of 120
10146704 Domed Strip Cap 8-tube Strips PCR Clean (Non-sterile) 0.1 mL Pack of 120
10167184 Flat Strip Cap 8-tube Strips PCR Clean (Non-sterile) 0.1 mL Pack of 120
15983982 Not Included Fast PCR Tube Strips PCR Clean (Non-sterile) 0.1 mL Pack of 120
15903992 Domed Strip Cap Fast PCR Tube Strips PCR Clean (Non-sterile) 0.1 mL Pack of 120
15993982 Flat Strip Cap Fast PCR Tube Strips PCR Clean (Non-sterile) 0.1 mL Pack of 120
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 15645367 Supplier Eppendorf™ Supplier No. 0030124707
Only null left
Add to basket
Edge
Add to basket
Eppendorf PCR Tubes ensure uniform heat distribution to the sample for optimal PCR performance. The user-friendly design allows for easy opening and provides a secure seal to prevent evaporation.
Ensure efficient and homogenous heat transfer with these thin-walled polypropylene tubes. Eppendorf™ Polypropylene PCR Tubes are easy to open, but provide tight sealing to prevent evaporation in PCR.
  • Design ensures an airtight seal to prevent contamination and evaboration.
  • Thin polypropylene walls ensure efficient and homogeneous heat transfer or fast and high-quality PCR results.
  • 8 reaction tubes in strip-format – ideal for small sample volumes. Easily adaptable for automation.
  • Strips are sealable using flat or domed strip-lids.
  • PCR Clean: Certified free from human DNA, DNase, RNase and PCR inhibitors).
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Volume (Metric) 0.2 mL
Quantity 500 Pk. (10 Bags of 50)
For Use With (Equipment) With all thermal cyclers with 0.2mL block format
Sterility Non-sterile
Material Polypropylene
Purity or Quality Grade Forensic DNA Grade
Closure Type Hinged Cap
Autoclavable Autoclavable
Format Single Tube
Product Type PCR Tube