Learn More
Fisherbrand™ Pneumatic Basic Crimping Tool
Reduces hand strain compared to manual crimper operation.
Brand: Fisherbrand™
2555.50 EUR valid until 2024-12-31
Use promo code "23262" to get your promotional price.
Additional Details : Weight : 0.80000kg
Description
- Includes pressure regulator
- Safety valve and nylon (PA) twisted-hose
- Pneumatic
- Steel
Specifications
One hand, secure, reproducible crimps/decrimps | |
8, 11, 13 and 20 mm vials | |
Pneumatic Hand-held Crimping Tool, pressure regulator, safety valve, nylon twisted hose |