Learn More
Fisherbrand™ Replacement Electrode Wire
Product Code. p-8007910 Shop All Fisherbrand Products
Change view
Click to view available options
Dimensions (L x W):
1m x 0.2 mm
500 x 0.2 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) unitSize
11817583 500 x 0.2 mm Each
11887803 1m x 0.2 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11817583
Only null left
Add to basket
Add to basket
Replacement Electrode Wire is an accessoty for use with Verti-Gel Maxi, 2-Gel and 4-Gel Electrophoresis System.
  • Made of platinum
Specifications
For Use With (Equipment) Major types of 80 x 100 mm and 100 x 100 mm Precast Gel
Type Electrode Wire Replacement
Material Platinum
Dimensions (L x W) 500 x 0.2 mm