Learn More
Thermo Scientific™ Pierce™ StartingBlock™ Blocking Buffers
Block membranes, plates and other samples with these ready-to-use PBS or TBS formulations of a special protein blocker for exceptional nonspecific blocking and broad compatibility.
42.95€ - 249.00€
Specifications
For Use With (Application) ELISA, WB
View More Specs
Products 6
Product Code Brand Product Name Formulation Quantity Price Quantity & Availability  
Product Code Brand Product Name Formulation Quantity Price Quantity & Availability  
10108313 Thermo Scientific™
37538
StartingBlock (PBS) Blocking Buffer Proprietary protein formulation in phosphate-buffered saline, pH 7.5 1L 236.00€
1 L View alternative products
Save Up to 
»
EU Stock 19
Add to basket
13424299 Thermo Scientific™
37579
StartingBlock (TBS) Blocking Buffer Proprietary protein formulation in Tris-buffered saline at pH 7.5 100mL 42.95€
100mL View alternative products
Save Up to 
»
EU Stock 3
Add to basket
13414299 Thermo Scientific™
37578
StartingBlock (PBS) Blocking Buffer Proprietary protein formulation in phosphate-buffered saline at pH 7.5 100mL 42.95€
100mL View alternative products
Save Up to 
»
EU Stock 4
Add to basket
10371804 Thermo Scientific™
37542
StartingBlock (TBS) Blocking Buffer Proprietary protein formulation in phosphate-buffered saline, pH 7.5 1L 236.00€
1 L View alternative products
Save Up to 
»
EU Stock 50
Add to basket
10381804 Thermo Scientific™
37543
StartingBlock T20 (TBS) Blocking Buffer Proprietary protein formulation in phosphate-buffered saline, pH 7.5, with 0.05% Tween 20 1L 249.00€
1 L View alternative products
Save Up to 
»
EU Stock 49
Add to basket
10270404 Thermo Scientific™
37539
StartingBlock T20 (PBS) Blocking Buffer Proprietary protein formulation in phosphate-buffered saline, pH 7.5, with 0.05% Tween 20 1L 249.00€
1 L View alternative products
Save Up to 
»
EU Stock 20
Add to basket
Description
Description
Although no blocking buffer is ideal for every experimental situation, you are likely to obtain excellent results from the start with StartingBlock Blocking Buffers. This is because the blocking agent is performs well with an extremely wide range of antibodies and assay systems. Our scientists have yet to encounter an antibody combination or other protein probing system in which StartingBlock Blocking Buffer is not compatible.
Highlights:
  • Compatible with many detection systems – Western blot, ELISA and IHC with antibody or avidin/biotin probes (blocker is serum and biotin-free)
  • Short blocking times – less than 15 minutes for nitrocellulose or PVDF membranes, almost instantaneous for polystyrene microplate wells
  • Strip and reprobe without reblocking – blots stay blocked even after stripping with Thermo Scientific Restore Stripping Buffer (Part No. 21059)
  • High signal:noise ratio – biotin- and serum protein-free formulation contributes to acheiving signal-to-noise ratios from 10:1 to 20:1
  • Convenient formats – available ready-to-use 1X formulations in PBS and TBS, with and without Tween™-20 detergent
Specifications
Specifications
ELISA, WB