Learn More
Thermo Scientific™ Pierce™ Protein-Free Blocking Buffers
Block membranes, plates and other samples with these ready-to-use PBS and TBS formulations of a non-protein compound to yield extremely low background.
42.95€ - 209.00€
Specifications
Applications ELISA, IHC, WB
View More Specs
Products 6
Product Code Brand Description Quantity Price Quantity & Availability  
Product Code Brand Description Quantity Price Quantity & Availability  
13474299 Thermo Scientific™
37584
Protein-Free (PBS) Blocking Buffer 100mL 44.05€
100mL View alternative products
Save Up to 
»
EU Stock 10
Add to basket
10147483 Thermo Scientific™
37573
Protein-Free T20 (PBS) Blocking Buffer 1L 209.00€
1 L View alternative products
Save Up to 
»
EU Stock 10
Add to basket
10137483 Thermo Scientific™
37571
Protein-Free T20 (TBS) Blocking Buffer 1L 209.00€
1 L View alternative products
Save Up to 
»
EU Stock 7
Add to basket
10269613 Thermo Scientific™
37570
Protein-Free (TBS) Blocking Buffer 1L 193.00€
1 L View alternative products
Save Up to 
»
EU Stock 25
Add to basket
13484299 Thermo Scientific™
37585
Protein-Free (TBS) Blocking Buffer 100mL 42.95€
100mL View alternative products
Save Up to 
»
EU Stock 5
Add to basket
10229663 Thermo Scientific™
37572
Protein-Free (PBS) Blocking Buffer 1L 193.00€
1 L View alternative products
Save Up to 
»
EU Stock 30
Add to basket
Description
Description
Traditional blocking buffers contain proteins that can cross-react with immunodetection systems, resulting in high background and reduced signal. Typical commercial alternatives to protein-based blocking agents are ineffective. Pierce Protein-Free Blocking Buffers are devoid of protein while remaining highly effective at blocking plates, membranes and other surfaces for ELISA, Western blotting, glass slide arrays and other applications.
Highlights:
  • Protein-free blocker – minimizes or eliminates cross-reactivity associated with protein-based blocking buffers
  • Application-compatible – effective in all kinds of protein detection systems, including Western blots (membranes), ELISA (microplates) and arrays (coated glass slides)
  • Streptavidin-friendly – absolutely free of biotin; no possible interference with avidin-biotin detection systems
  • High-performance – optimized and validated in many protein methods to provide high signal-to-noise ratio (i.e., no quenching of specific binding and signal but eliminating nonspecific binding and background)
  • Convenient – supplied as ready-to-use 1X formulations in TBS and PBS with and without 0.05% Tween™-20
Specifications
Specifications
ELISA, IHC, WB