Learn More
Thermo Scientific™ Pierce™ Protease Inhibitor Tablets, EDTA-free
Product Code. p-7078347 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
20 Tablets
Unit Size:
Pack of 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15677308 20 Tablets Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
For Research Use Only. Not for use in diagnostic procedures.
Product Code. 15677308 Supplier Thermo Scientific™ Supplier No. A32965
For Research Use Only. Not for use in diagnostic procedures.
Thermo Scientific™ Pierce Protease Inhibitor Tablets are broad-spectrum protease inhibitors that are highly effective at preventing proteolytic degradation for cell lysis and protein extraction experiments
Pierce Protease Inhibitor Tablets protect proteins from proteolytic degradation through inhibition of the serine-proteases, cysteine-proteases, aspartic acid-proteases and aminopeptidases that are typically present in cellular lysate samples. The tablets contain AEBSF, aprotinin, bestatin, E-64, leupeptin and pepstatin A. Stable for storage at 4°C for up to 12 months, each mini or large tablet tablet is sufficient for 10mL or 50mL of solution, respectively, and is suitable for reagent preparation prior to protein extraction from tissue and cultured cells.
Highlights:
  • Improved formulation&mdsashtablets dissolve quickly into a clear solution
  • Economical – more cost-effective than other commercially available formulations for equivalent volumes
  • Convenient&mdsashready-to-use, fully disclosed, broad-spectrum formulations
  • Compatible&mdsashuse directly with Thermo Scientific™ Pierce™ BCA Assays
  • EDTA-free- formulated without EDTA, a metalloproteinase inhibitor (only for EDTA-free formulations)
  • TRUSTED_SUSTAINABILITY
    Specifications
    Product Type Protease Inhibitor Cocktail
    Content And Storage 1 Vial, Store at 2°C to 8°C
    Shipping Condition Room Temperature
    Form Solid
    Product Line Pierce
    Sufficient For 50 mL Solution
    Inhibitors Protease Inhibitor
    Quantity 20 Tablets
    compliance-icons
    Product Identifier
    • Pierce Protease Inhibitor Tablets, EDTA-Free
    Signal Word
    • Danger
    Hazard Category
    • Serious eye damage/eye irritation Category 1
    • Skin corrosion/irritation Category 1 B
    Hazard Statement
    • H314-Causes severe skin burns and eye damage.
    • H318-Causes serious eye damage.
    Precautionary Statement
    • P264-Wash hands thoroughly after handling.
    • P280-Wear protective gloves/protective clothing/eye protection/face protection.
    • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
    • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
    • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
    • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    • P310-Immediately call a POISON CENTER/doctor/.
    • P330-Rinse mouth.
    • P331-Do NOT induce vomiting.
    • P363-Wash contaminated clothing before reuse.
    Supplemental information
    • MIXTURE LIST-Contain: 4-(2-aminoethyl)-Benzenesulfonyl fluoride hydrochloride