Learn More
Thermo Scientific™ Pierce™ PNPP Substrates
Powders, tablets, solutions and complete kits of p-nitrophenyl phosphate, a colorimetric, soluble alkaline phosphatase substrate for use in ELISA applications.
157.00€ - 204.00€
Products 3
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
10571384 Thermo Scientific™
37620
Pierce PNPP Substrate Kit 204.00€
1 set View alternative products
Save Up to 
»
EU Stock 12
Add to basket
11889260 Thermo Scientific™
34045
Pierce PNPP Powder 157.00€
25g View alternative products
Save Up to 
»
EU Stock 3
Add to basket
11859270 Thermo Scientific™
34047
Pierce PNPP Tablets 160.00€
105 tablets View alternative products
Save Up to 
»
EU Stock 16
Add to basket
Description
Description
PNPP (p-nitrophenyl phosphate, disodium salt) is a widely used substrate for detecting alkaline phosphatase in ELISA applications. When alkaline phosphatase and PNPP are reacted, a yellow water-soluble reaction product is formed. This reaction product absorbs light at 405nm. Pierce PNPP is available in four formats: dry crystalline powder, single-use tablets, a stable substrate solution and a complete kit containing substrate solution and diethanolamine buffer.
Highlights:
  • Absorbance maximum of 505nm for measurement by ELISA plate readers
  • Develops a yellow hue in solution upon reaction with alkaline phosphatase (AP)
  • Safer to use – solution contains a non-mercury, non-azide preservative