missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ ABC Staining Kits

Detect biotinylated secondary antibodies by IHC using the highly sensitive avidin-biotin complex (ABC) technique.

334.00€ - 622.00€

Specifications

Detection Chromogenic
Kit Contents

Blocking Serum (horse), 3mL

,

Biotinylated Horse Anti-Mouse IgG, 1mL

,

Reagent B (Biotinylated HRP). 2mL

,

Reagent A (Avidin), 2mL

View More Specs

Products 4
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
10485695 Thermo Scientific™
32052
Pierce Ultrasensitive ABC Peroxidase Mouse IgG Staining Kit 622.00€
1 set View alternative products
Save Up to 
»
EU Stock 2
Add to basket
 
10435685 Thermo Scientific™
32020

Pierce ABC Peroxidase Standard Staining Kit

334.00€
1 set View alternative products
Save Up to 
»
EU Stock 14
Add to basket
 
10570814 Thermo Scientific™
32054
Pierce Ultrasensitive ABC Peroxidase Rabbit IgG Staining Kit 618.00€
1 set View alternative products
Save Up to 
»
EU Stock 1
Add to basket
 
10188013 Thermo Scientific™
32050
Pierce Ultrasensitive ABC Standard Peroxidase Staining Kit 436.00€
1 set View alternative products
Save Up to 
»
EU Stock 13
Add to basket
 
Description

Description

Highly diluted primary antibodies can be used with ABC Staining Kits, producing comparable stain intensity to other methods that require higher concentrations of antibody. Kits with a secondary antibody are selected by the species of primary antibody to be used. For example, if the primary antibody (IgG) is produced in mice, select the ABC Mouse IgG Kit. All you need to create a sensitive detection system is your specific primary antibody, an ABC Kit with biotinylated antibody and a substrate for horseradish peroxidase.The Standard Kits include only the avidin and biotinylated enzyme. They are useful if an ABC Kit is not available for your specific species of primary antibody, or if the primary antibody is directly labeled with a Thermo Scientific EZ-Link Biotinylation Reagent. The biotinylated antibody for your specific application can then be used, along with blocking serum and a Standard ABC Kit.

Highlights:

  • Produce very low background staining
  • Require lower primary antibody concentrations than standard methods
  • Ideal for immunohistochemistry, ELISA, and Western blotting applications

ABC Kits

  • Available with horseradish peroxidase
  • Contain a biotinylated secondary antibody in a choice of species – the species should be selected based on the species of the primary antibody to be used

Standard Kits

  • Available with a choice of alkaline phosphatase or horseradish peroxidase
  • Suitable for use when an ABC kit is not available for the species of the primary antibody or if the specificity of the antibody in the kit is not appropriate for the application
  • Do not include secondary antibody or blocking serum – secondary antibody and blocking serum appropriate to the application should be purchased separately

Ultrasensitive ABC Kits

  • Available with horseradish peroxidase
  • Provide greater sensitivity than ABC Kits without increased background staining
  • Supply the extra sensitivity needed for localizing antigens present in very small quantities
  • Permit approximately five-fold higher dilutions of primary antibody to be used than do ABC Kits

Includes:

Biotinylated affinity-purified secondary antibody, 1mL (except Standard Kits); Appropriate blocking serum, 3mL (except Standard Kits); Avidin reagent, 2mL; Biotinylated enzyme reagent, 2mL

Specifications

Specifications

Chromogenic

Blocking Serum (horse), 3mL

,

Biotinylated Horse Anti-Mouse IgG, 1mL

,

Reagent B (Biotinylated HRP). 2mL

,

Reagent A (Avidin), 2mL