missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Phosphatase, Alkaline, Calf Intestine, Protein
133.00€ - 423.00€
Specifications
Concentration | Concentration on Label |
---|---|
Formulation | In 5mM MgCl2, 5mM Tris-HCl, 100μM ZnCl2, 50% glycerol, pH 7.0. |
Preparation Method | Dilute buffer or H2O |
Immunogen | Phosphatase, Alkaline |
Regulatory Status | RUO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17669895
|
Merck Chemicals
524572-5KU |
5kU |
423.00€
5ku |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||||
17659895
|
Merck Chemicals
524572-1KU |
1kU |
133.00€
1ku |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||||
Description
Native, alkaline phosphatase from calf intestine. Homodimeric enzyme that hydrolyzes phosphate esters of primary and secondary alcohols, amines, and phenols. Useful in hydrolysis of 5'-terminal phosphates of DNA and RNA. Note: 1KU = 1,000 units.Specifications
Concentration on Label | |
Dilute buffer or H2O | |
RUO | |
Native |
In 5mM MgCl2, 5mM Tris-HCl, 100μM ZnCl2, 50% glycerol, pH 7.0. | |
Phosphatase, Alkaline | |
Bovine | |
Liquid |