Learn More
Thermo Scientific™ Owl™ S3S System Combs
Eliminate comb assembly with these heavy-duty, one-piece combs.
55.16€ - 63.60€
Specifications
For Use With (Equipment) | Owl™ S3S Aluminum-Backed Sequencer System |
---|---|
Product Type | Combs |
Product Code | Brand | Description | Teeth Count | Thickness (Metric) Teeth | Volume (Metric) Well | Price | Quantità e disponibilità | ||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Description | Teeth Count | Thickness (Metric) Teeth | Volume (Metric) Well | Prezzo | Quantità e disponibilità | ||
12444717
|
Thermo Scientific™
S2S-MT2 |
Sharktooth Micro-Well Comb | 72 (2X) | 0.4 mm | 10µL |
63.60€
Each |
In Stock
Log in to see stock. |
||
12454717
|
Thermo Scientific™
S2S-MT3 |
Sharktooth Comb | 110 (3X) | 0.4 mm | 5.5µL |
63.60€
Each |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||
12474717
|
Thermo Scientific™
S2S-MT44 |
Sharktooth Micro-Well Comb | 146 (4X) | 0.4 mm | 3.7µL |
63.60€
Each |
Estimated Shipment: 22-08-2024
Log in to see stock. |
||
11505195
|
Thermo Scientific™
S2S-MT42 |
Sharktooth Micro-Well Comb | 146 (4X) | 0.2 mm | 2µL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||
Descrizione
Eliminate comb assembly with Thermo Scientific™ Owl™ S3S System Combs. These heavy-duty, one-piece combs are for use with the Owl™ S3S System.
Heavy duty, one piece combs eliminate comb assembly.Specifica
Owl™ S3S Aluminum-Backed Sequencer System | |
Combs |