Learn More
Novus Biologicals™ Imidazoquinoline Resiquimod (R-848), TLR7 and TLR8 Ligand
Applications: Functional
Brand: Novus Biologicals™ NBP2-26231
Additional Details : Weight : 0.00970kg
Description
Imidazoquinoline Resiquimod (R-848), TLR7 and TLR8 ligand is a low molecular weight synthetic molecule of the Imidazoquinoline family. It has potent immunostimulatory activity. It stimulates antibody secretion, cytokine production, protection from apoptosis and CD80 up-regulation.
- Species: Human
- Stimulation of TLR8 has been achieved with 0.5-5µg/mL
Recommended Storage
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Specifications
DMSO | |
Identity determined by MS, 1H-NMR and 13C-NMR. This product is useful for activation of mouse TLR7 and human TLR7 and TLR8. Stimulation of TLR8 has been achieved with 0.5-5 ug/mL. Use in functional assays reported in scientific literature (PMID 26134251). | |
>98%, by HPLC. |
Store at -20°C. | |
Imidazoquinoline Resiquimod (R-848), TLR7 and TLR8 Ligand | |
0.5 mg |