missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Neurokinin A
For cellular and molecular biology applications
60.50€ - 86.10€
Specifications
Purification Method | HPLC |
---|---|
Common Name | Substance K |
Content And Storage | -30°C to -10°C |
Format | Powder |
Molecular Weight (g/mol) | 1133.33 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15438329
|
Thermo Scientific Chemicals
J66817.!D |
0.5 mg |
60.50€
0.50mg |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
15448329
|
Thermo Scientific™
J66817.M^ |
1 mg |
86.10€
1mg |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
HPLC | |
-30°C to -10°C | |
1133.33 | |
RUO - research use only |
Substance K | |
Powder | |
Neurokinin A | |
His-Lys-Thr-Asp-Ser-Phe-Val-Gly-Leu-Met-NH2 |
RUO – Research Use Only