Learn More
N-sulfated Heparan Sulfate Monoclonal Antibody (HepSS-1), Biotin, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA546186
Additional Details : Weight : 0.01000kg
Description
Heparan sulfate (HS) is a glycosaminoglycan that is ubiquitously expressed on cell surfaces and in the extracellular matrix and basement membrane. Each HS molecule is a linear polysaccharide composed of repeating disaccharides of hexuronic acid and d-glucosamine that can exhibit immense structural diversity due to substitution to varying extents with sulfate groups and epimerization of glucuronic acid to iduronic acid, with areas of high sulfation and glucuronic acid epimerization being co-located in hot spots throughout the molecule. HS is structurally related to heparin, an extremely highly sulfated form of HS that is restricted to mast cells. The biosynthesis and modification of HS chains is thought to take place within the endoplasmic reticulum, Golgi apparatus, and trans Golgi network, which in the end produce unique HS chains that are covalently attached to a range of core proteins to form HS-proteoglycans [PMID: 24391644].Specifications
N-sulfated Heparan Sulfate | |
Monoclonal | |
1 mg/mL | |
TBS with 15mM sodium azide; pH 8 | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
Chemical | |
Liquid | |
Chemical |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
HepSS-1 | |
Biotin | |
Heparan sulfate | |
MethA murine fibrosarcoma | |
100 μg | |
Primary | |
4° C, do not freeze | |
IgM |