Learn More
3M™ Mustard Protein ELISA Kit, E96MUS
Intended for the detection of mustard proteins in clean-in-place water (CIP) final rinse water, environmental swab samples, food ingredients and processed food products.
Brand:  3M™ 7100151337
Additional Details : Weight : 0.24900kg
Product Code. 17253804
  • 899.94€ / 1 set
Estimated Shipment: 07-08-2024
to see stock.
Add to basket
Beschreibung
Beschreibung
  • Designed to detect both processed and unprocessed target allergen proteins.
  • Test can accommodate environmental swab, CIP rinse water and food samples.
  • A universal extraction protocol streamlines testing for multiple analytes, saving you reagents and valuable time.
Spezifikation
Spezifikation
Test
96 Wells
50 Minutes
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Sonderangebote
Sonderangebote