Learn More
3M™ Mustard Protein ELISA Kit, E96MUS
Intended for the detection of mustard proteins in clean-in-place water (CIP) final rinse water, environmental swab samples, food ingredients and processed food products.
Brand: 3M™ 7100151337
Additional Details : Weight : 0.24900kg
Beschreibung
- Designed to detect both processed and unprocessed target allergen proteins.
- Test can accommodate environmental swab, CIP rinse water and food samples.
- A universal extraction protocol streamlines testing for multiple analytes, saving you reagents and valuable time.
Spezifikation
Test | |
96 Wells | |
50 Minutes |