Learn More
MP Biomedicals™ γ-Globulin, Bovine Purified
Suitable for use as a carrier protein in secondary antibody systems, e.g., radioimmunoassay. Purified proteins are recommended as ELISA coating antigens and as controls.
Brand:  MP Biomedicals™ 0855847
Additional Details : Weight : 0.00010kg
Product Code. 17592936
  • 501.38€ / 100mg
Estimated Shipment: 04-07-2024
Specifications
20 mg/mL
2°C to 8°C
Immunoassay
100 mg
Lyophilized
Purified Bovine gamma Globulin