Learn More
MP Biomedicals™ Freund's Complete Adjuvant
Brand:  MP Biomedicals™ 0855828
160.69 EUR valid until 2024-12-31
Use promo code "23257" to get your promotional price.
Additional Details : Weight : 0.05000kg
Product Code. 11424225
  • 171.72€ / 50mL
Estimated Shipment: 04-07-2024
Specifications
2°C to 8°C
Used to produce water-in-oil emulsions of immunogens, Immunoassays
50 mL
Freund's Complete Adjuvant
Paraffin oil 80 to 90%, Arlacel A 10 to 20%, heat-killed Mycobacterium butyricum 0.1 to 1.5%