missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IgM Isotype Control (PFR-03), Invitrogen™
Mouse Isotype Control
Brand: Invitrogen MA110438
Additional Details : Weight : 0.01000kg
Description
MA1-10438 can be used as a negative control to measure the non-specific binding of Mouse IgM in human, porcine, mouse and rat tissues.
Mouse IgM is an immunoglobulin M antibody that is produced by B cells in response to an antigen. It is the first antibody produced during an immune response and is the largest immunoglobulin in the serum. Mouse IgM is composed of five heavy chains and five light chains, and it is involved in complement activation and agglutination of antigens. It plays a crucial role in the innate immune response and is important for neutralizing pathogens before the adaptive immune response is fully activated. Mouse IgM is widely used in research and diagnostic applications, including ELISA, Western blotting, and immunohistochemistry.Specifications
Mouse IgM | |
PFR-03 | |
Unconjugated | |
AGM1; AI326478; Igh6; Igh-6; Ighm; Igh-M; Igm; immunoglobulin heavy constant mu; MU; muH; VH | |
Phytophthora sp. | |
100 μg | |
Isotype Control (Primary) | |
4° C, do not freeze | |
IgM |
Control, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation | |
1 mg/mL | |
TBS with 15mM sodium azide; pH 8.0 | |
Mouse | |
sequential chromatography | |
RUO | |
Not Applicable | |
Liquid |