Learn More
Mouse IgG1 Isotype Control (MOPC-21), Invitrogen™
Mouse Isotype Control
Marca: Invitrogen MA110407
Detalles adicionales : Weight : 0.01000kg
Descripción
MA1-10407 can be used as a negative control to measure the non-specific binding of Mouse IgG1 in human and rat tissues. Endotoxin level is less than 0.01 EU/ug of the protein, as determined by the LAL test.
Mouse IgG1 is a subtype of immunoglobulin G antibody that is produced by B cells in response to an antigen. It is the most abundant IgG subtype in the serum and plays a crucial role in the adaptive immune response. Mouse IgG1 is composed of two heavy chains and two light chains, and it is involved in opsonization, complement activation, and antibody-dependent cell-mediated cytotoxicity. It is also involved in the regulation of immune responses, including the activation of T cells and the inhibition of B cell activation. Mouse IgG1 is widely used in research and diagnostic applications, including ELISA, Western blotting, and immunohistochemistry. It is also used in therapeutic applications, such as immunotherapy and vaccine development.Especificaciones
Mouse IgG1 | |
MOPC-21 | |
Unconjugated | |
IgG1; Igh-4; Ighg1; Immunoglobulin G; Immunoglobulin G1; immunoglobulin heavy constant gamma 1 (G1m marker); ImmunoglobulinG; ImmunoglobulinG1; VH7183 | |
not available | |
100 μg | |
Isotype Control (Primary) | |
4° C, do not freeze | |
IgG1 κ |
Control, Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with no preservative; pH 7.4 | |
Mouse | |
Protein A | |
RUO | |
Not Applicable | |
Liquid |