missing translation for 'onlineSavingsMsg'
Learn More

Mouse IgG1 Isotype Control (MOPC-21), DyLight™ 650, Invitrogen™

Mouse Isotype Control

Brand:  Invitrogen MA1191D650

Additional Details : Weight : 0.00900kg

Product Code. 15547211

  • 372.00€ / 100µg
Estimated Shipment: 18-06-2024
to see stock.
Add to basket

Description

Description

MA1-191-D650 can be used as a negative control in any immunoassay where a DyLight 650-conjugated primary antibody of the mouse IgG1 isotype is used. DyLight 650 has an excitation/emission of 652/672nm.

Mouse IgG1 Control antibody specifically detects Mouse IgG1 in Not Applicable samples. It is validated for Control, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.
Specifications

Specifications

Mouse IgG1
MOPC-21
DyLight 650
IgG1; Igh-4; Ighg1; Immunoglobulin G; Immunoglobulin G1; immunoglobulin heavy constant gamma 1 (G1m marker); ImmunoglobulinG; ImmunoglobulinG1; VH7183
Size-exclusion chromatography
RUO
Not Applicable
Liquid
Control, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
0.1 mg/mL
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4
Mouse
100 μg
Isotype Control (Primary)
4° C, do not freeze
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers