Learn More
Eppendorf™ Centrifuge 5430/5430 R
The unique, cross-over Eppendorf Centrifuge 5430 and 5430 R combine the small footprint of a microcentrifuge with the versatility of a multipurpose centrifuge. Extremely compact, they can accommodate rotors for microplates and 15/50 mL conical tubes as well as Eppendorf tubes and PCR strips.
3820.00€ - 8150.00€
Specifications
Display | Large, brightly lit LCD |
---|---|
Safety Features | Lid Lock, steel armoured safety chamber, view port for speed validation, lid emergency release, automatic rotor recognition, certified aerosol-tight rotor lids, automatic imbalance recognition |
Timer | 30 sec. to 99:59 hrs., with continuous run function |
Certifications/Compliance | UL (120 V), CAN/CSA (120 V), CE, RoHS, IVD for certain catalog numbers |
Frequency | 50/60 Hz |
Product Code | Brand | Model | Refrigerated | Includes | Electrical Requirements | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Refrigerated | Includes | Electrical Requirements | Price | Quantity & Availability | ||
10326924
|
Eppendorf™
5427000410 |
Centrifuge 5430, rotary knobs | No | With Rotor FA-45-30-11 incl. rotor lid | 230 V, 50/60 Hz (EU) |
4520.00€
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||
10091594
|
Eppendorf™
5427000615 |
Centrifuge 5430, rotary knobs | No | Without rotor | 230 V, 50/60 Hz (EU) |
4010.00€
Each |
In Stock
Log in to see stock. |
||
10329443
|
Eppendorf™
5428000619 |
Centrifuge 5430 R, rotary knobs | Yes | Without rotor | 230 V/50-60 Hz (EU) |
8150.00€
Each |
In Stock
Log in to see stock. |
||
15825073
|
Eppendorf™
5428000010 |
Centrifuge 5430 R, keypad | Yes | With Rotor FA-45-30-11 incl. rotor lid | 230 V, 50/60 Hz (EU) |
8055.00€
1 set |
In Stock
Log in to see stock. |
||
15845073
|
Eppendorf™
5428000205 |
Centrifuge 5430 R, keypad | Yes | Without rotor | 230 V, 50/60 Hz (EU) |
7765.00€
Each |
|
||
15865073
|
Eppendorf™
5428000410 |
Centrifuge 5430 R, rotary knobs | Yes | With Rotor FA-45-30-11 incl. rotor lid | 230 V, 50/60 Hz (EU) |
8055.00€
1 set |
In Stock
Log in to see stock. |
||
15885073
|
Eppendorf™
5428000610 |
Centrifuge 5430 R, rotary knobs | Yes | Without rotor | 230 V, 50/60 Hz (EU) |
7765.00€
Each |
In Stock
Log in to see stock. |
||
15865063
|
Eppendorf™
5427000210 |
Centrifuge 5430, keypad | No | With Rotor FA-45-30-11 incl. rotor lid | 230 V, 50/60 Hz (EU) |
4305.00€
1 set |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||
15845063
|
Eppendorf™
5427000015 |
Centrifuge 5430, keypad | No | Without rotor | 230 V, 50/60 Hz (EU) |
3820.00€
Each |
In Stock
Log in to see stock. |
||
16692832
|
Eppendorf™
5427000178 |
Centrifuge 5430, rotary knobs | No | With Rotor A-2-MTP | 230 V, 50/60 Hz (EU) |
5015.00€
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||
15885063
|
Eppendorf™
5427000415 |
Centrifuge 5430, rotary knobs | No | With Rotor FA-45-30-11 incl. rotor lid | 230 V, 50/60 Hz (EU) |
4305.00€
1 set |
Estimated Shipment: 12-06-2024
Log in to see stock. |
||
15805073
|
Eppendorf™
5427000610 |
Centrifuge 5430, rotary knobs | No | Without rotor | 230 V, 50/60 Hz (EU) |
3820.00€
Each |
|
||
Description
Specifications
Large, brightly lit LCD | |
30 sec. to 99:59 hrs., with continuous run function | |
50/60 Hz |
Lid Lock, steel armoured safety chamber, view port for speed validation, lid emergency release, automatic rotor recognition, certified aerosol-tight rotor lids, automatic imbalance recognition | |
UL (120 V), CAN/CSA (120 V), CE, RoHS, IVD for certain catalog numbers | |
230 V |