Learn More
Novacyt - Microgen Bioproducts Latex Agglutination Test: Salmonella
Can be used to obtain rapid, presumptive-positive results from selective media for targeted identification/diagnosis.
Brand: Novacyt - Microgen Bioproducts M42CE
Additional Details : Weight : 0.90000kg
Description
- From a range of latex products for the rapid diagnosis of the most common pathogens for foodborne illness.
- Specific tests available for a variety of infection detections.
- Rapid (2 minute), simple, sensitive, and specific latex agglutination test for the identification and confirmation of infecting colonies.
- Can be used to obtain rapid, presumptive-positive results and provide an early indication of possible production process failure.
- Can be used to screen colonies before full biochemical identification and can replace traditional agglutinating antibody methods.
Specifications
2°C to 8°C | |
Latex Agglutination Test | |
2 min. |
Salmonella | |
50 Tests |