Learn More
OHAUS™ Large Two Prong Sleeves
OHAUS™ Large Two Prong Sleeves are for use with clamps and can be easily removed for cleaning or replacement.
9.55€
Product Code | Brand | Material | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material | Price | Quantity & Availability | |||||
15742669
|
OHAUS™
30400255 |
Vinyl |
9.55€
Each |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
15752669
|
OHAUS™
30400256 |
Fiberglass |
9.55€
Each |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
Description
- Available in red vinyl and fiberglass
- Fiberglass sleeves are ideal for application above 100°C
Specifications
2 | |
Prong Sleeve | |
Large |