missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Lab Vision™ UltraVision™ Large Volume Detection System: anti-Polyvalent, HRP (Ready-To-Use)

A versatile and robust biotin-streptavidin detection system for research applications and cost-conscious laboratories.

485.00€ - 880.00€

Specifications

Chromagen None provided
Specificity anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)
Enzyme Peroxidase
Species Goat
Storage Requirements Store at 2-8°C. Each component is stable for 18 months.
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
12684047 Thermo Scientific
TP-060-HL
60mL 485.00€
60mL View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 30-05-2019
Add to basket
 
12634057 Thermo Scientific™
TP-125-HL
125mL 880.00€
125mL View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 30-05-2019
Add to basket
 
Description

Description

This technique involves the sequential incubation of the specimen with an unconjugated primary antibody specific to the target antigen, a biotinylated secondary antibody that reacts with the primary antibody, enzyme-labeled streptavidin, and substrate/chromogen.
  • Labeled streptavidin-biotin immunoenzymatic antigen detection system
  • Species: Goat

  • Specificity: anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)

  • Enzyme: Peroxidase

  • Chromogen/Substrate: None provided

Includes:

TP-060-HL: 60mL Ultra V Block (TA-060-UB), 60mL Biotinylated Goat Anti-Polyvalent (TP-060-BN), 60mL Streptavidin Peroxidase (TS-060-HR)

TP-125-HL: 125mL Ultra V Block (TA-125-UB), 125mL Biotinylated Goat Anti-Polyvalent (TP-125-BN), 125mL Streptavidin Peroxidase (TS-125-HR)

Note:

UltraVision detection system detects a specific antibody bound to an antigen in tissue sections. The specific antibody is located by a biotin-conjugated secondary antibody. This step is followed by the addition of a streptavidin-enzyme conjugate that binds to the biotin present on the secondary antibody. The specific antibody, secondary antibody, and streptavidin-enzyme complex is then visualized with an appropriate substrate/chromogen.

Specifications

Specifications

None provided
Peroxidase
Store at 2-8°C. Each component is stable for 18 months.
anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)
Goat
None provided