Learn More
DWK Life Sciences Kimble™ Kontes™ Chromatography Column PEEK Unions
Tubing nuts and ferrules are used to make connections between the tubing and the end fittings of CHROMAFLEX™, FLEX-COLUMN™, DISPOSAFLEX™ columns, and ULTRA-WARE™ filtration assemblies.
Brand:  DWK Life Sciences 420818-3305
Additional Details : Weight : 1.00000kg
Product Code. 12913521
  • 79.45€ / Each
Estimated Shipment: 10-09-2024
Description
  • EFTE Flangeless Ferrule for 1.8mm OD Tubing
  • for Use with 420821 Series Male Nuts
  • CHROMAFLEX™ FITTINGS
  • Union connects tubing and end fittings F1/4-28 to F1/4-28
  • PEEK Internally Threaded Union for use with flangeless fittings
  • Case of 1
  • Replacement Parts in ULTRA-WARE™ Standard Fittings Kit
Specifications
Specifications
Chromatography Union
Flangeless fitting
PEEK
F1/4-28 to F5/16-24