Learn More
Thermo Scientific™ KingFisher™ Plastics for 96 standard and PCR formats
Optimize your performance using Thermo Scientific™ KingFisher consumables designed specifically for use with the KingFisher Flex, Duo Prime, and Presto instruments.
168.00€ - 482.00€
Product Code | Brand | Description | For Use With (Equipment) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | For Use With (Equipment) | Price | Quantity & Availability | ||||
16532452
|
Thermo Scientific™
97002560 |
KingFisher Apex 96 PCR tip comb (for Apex only) | KingFisher Apex only |
321.00€
Pack of 50 |
In Stock
Log in to see stock. |
||||
10357939
|
Thermo Scientific™
97002540 |
KingFisher 96 microplate (200μL) (for Flex and Presto) *Note: Only works with KingFisher™ Flex 96 Heating Block (Cat. No. 24075420). Not to be used with KingFisher™ Flex 96 Deep-Well Heating Block (Cat. No. 24075430). |
KingFisher Flex and Presto |
168.00€
Pack of 48 |
In Stock
Log in to see stock. |
||||
12613126
|
Thermo Scientific™
97002524 |
KingFisher 96 tip comb for KingFisher magnets, 100 pcs (for Flex) | KingFisher Flex |
468.00€
Pack of 100 |
In Stock
Log in to see stock. |
||||
12603126
|
Thermo Scientific™
97002514 |
KingFisher 96 tip comb for KingFisher PCR magnets, 80 pcs (for Flex only) | KingFisher Flex only |
378.00€
Pack of 80 |
Estimated Shipment: 27-05-2024
Log in to see stock. |
||||
17160069
|
Thermo Scientific™
97002524B |
KingFisher 96 Tip Comb, Barcoded | KingFisher Apex |
482.00€
Pack of 100 |
In Stock
Log in to see stock. |
||||
17180069
|
Thermo Scientific™
97002540B |
KingFisher 96 Plate (200 μL), Barcoded | KingFisher Apex |
186.00€
Pack of 48 |
In Stock
Log in to see stock. |
||||
Description
Optimize your performance using Thermo Scientific™ KingFisher Plastics designed specifically for use with the KingFisher Flex, Duo Prime, Presto and Apex instruments. KingFisher instruments utilize disposable plastic plates and tip combs made of polypropylene, which are ideal for magnetic particle processing due to their low binding affinity for biomolecules. Both the yield and quality of the isolated protein or nucleic acid are significantly improved with special plates and tip combs designed for KingFisher systems.
Use With: Thermo Scientific™ KingFisher instruments, including KingFisher Apex, KingFisher Flex and KingFisher Presto.Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.