Learn More
DWK Life Sciences Kimble™ Kontes™ Duall™ Tissue Grinders
Combined conical and cylindrical surfaces improve grinding efficiency
208.00€ - 263.00€
Specifications
For Use With (Application) | Ideal for connective tissue such as muscle, heart and lung |
---|---|
Clearance (English) | 0.004 to 0.006 in. |
Clearance (Metric) | 0.1 to 0.15mm |
Product Code | Brand | Capacity | Includes | Size | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Includes | Size | Price | Quantity & Availability | |||
11854005
|
DWK Life Sciences
885450-0020 |
0.03 oz., 1mL | Ground Glass pestle and tube | 20 |
263.00€
Each |
Estimated Shipment: 13-09-2024
Log in to see stock. |
|||
10545115
|
DWK Life Sciences
885450-0021 |
0.1 oz., 3mL | Ground Glass pestle and tube | 21 |
208.00€
Each |
Estimated Shipment: 13-09-2024
Log in to see stock. |
|||
Description
Combined conical and cylindrical surfaces improve grinding efficiency
- Can be motor driven
- Choose from all-ground glass style or smooth glass tube with PTFE pestle threaded to stainless-steel shaft
- Interchangeable tubes and pestles have 0.004 to 0.006 in. (0.10 to 0.15mm) clearance
Specifications
Ideal for connective tissue such as muscle, heart and lung | |
0.004 to 0.006 in. | |
0.1 to 0.15mm |