Learn More
Thermo Scientific™ KingFisher™ Plastics for 24 deep-well format
Optimize your performance using Thermo Scientific™ KingFisher consumables designed specifically for use with the KingFisher Flex, Duo Prime, and Presto instruments.
176.00€ - 844.00€
Product Code | Brand | Description | For Use With (Equipment) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | For Use With (Equipment) | Price | Quantity & Availability | ||||
16552452
|
Thermo Scientific™
97002580 |
KingFisher Apex 24 Combi tip comb (for Apex only) | For Use With KingFisher Apex only |
844.00€
Pack of 50 |
In Stock
Log in to see stock. |
||||
12653116
|
Thermo Scientific™
95040470 |
KingFisher 24 deep-well plate (for Duo Prime, Flex and Presto) | KingFisher Duo Prime, Flex and Presto |
216.00€
Pack of 50 |
In Stock
Log in to see stock. |
||||
12663116
|
Thermo Scientific™
95040480 |
KingFisher 24 deep-well plate, sterile, 50 pcs (for Duo Prime, Flex and Presto) | KingFisher Duo Prime, Flex and Presto |
268.00€
Pack of 50 |
In Stock
Log in to see stock. |
||||
12624075
|
Thermo Scientific™
97002610 |
KingFisher 24 deep-well tip comb and plate (for Flex and Presto) | KingFisher Flex and Presto |
630.00€
Pack of 50 |
In Stock
Log in to see stock. |
||||
12643116
|
Thermo Scientific™
97003510 |
KingFisher Duo Prime 6-tip comb and 24 deep-well plate (12 pieces of 24 deep-well plates, each including 4 tip combs)(for Duo Prime only) | For Use With KingFisher Duo Prime only |
176.00€
Pack of 48 |
In Stock
Log in to see stock. |
||||
17140069
|
Thermo Scientific™
95040470B |
KingFisher 24 Deep-well Plate, Barcoded | KingFisher Apex |
224.00€
Pack of 50 |
In Stock
Log in to see stock. |
||||
17190069
|
Thermo Scientific™
97002610B |
KingFisher 24 Deep-Well Tip Comb & Plate, Barcoded | KingFisher Apex |
634.00€
Pack of 50 |
Estimated Shipment: 17-06-2024
Log in to see stock. |
||||
17150069
|
Thermo Scientific™
95040480B |
KingFisher 24 Deep-Well Plate, Sterile, Barcoded | KingFisher Apex |
282.00€
Pack of 50 |
In Stock
Log in to see stock. |
||||
17100079
|
Thermo Scientific™
97002620B |
KingFisher 24 Deep-Well Tip Comb & Plate, Sterile, Barcoded | KingFisher Apex |
654.00€
Pack of 50 |
Estimated Shipment: 17-06-2024
Log in to see stock. |
||||
Description
Optimize your performance using Thermo Scientific™ KingFisher Plastics designed specifically for use with the KingFisher Flex, Duo Prime, Presto and Apex instruments. KingFisher instruments utilize disposable plastic plates and tip combs made of polypropylene, which are ideal for magnetic particle processing due to their low binding affinity for biomolecules. Both the yield and quality of the isolated protein or nucleic acid are significantly improved with special plates and tip combs designed for KingFisher systems.
Use With: Thermo Scientific™ KingFisher instruments, including KingFisher Flex, KingFisher Duo Prime, and KingFisher Presto.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.